5-Chlorobenzo[b]thiophene-3-acetic acid, 97%, Thermo Scientific Chemicals
5-Chlorobenzo[b]thiophene-3-acetic acid, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

5-Chlorobenzo[b]thiophene-3-acetic acid, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number L12836.MD
also known as L12836-MD
Price (USD)/ Each
72.30
-
Add to cart
Quantity:
250 mg
Request bulk or custom format
Price (USD)/ Each
72.30
Add to cart
5-Chlorobenzo[b]thiophene-3-acetic acid, 97%, Thermo Scientific Chemicals
Catalog numberL12836.MD
Price (USD)/ Each
72.30
-
Add to cart
Chemical Identifiers
CAS17266-30-7
IUPAC Name2-(5-chloro-1-benzothiophen-3-yl)acetic acid
Molecular FormulaC10H7ClO2S
InChI KeyQQKKTOPRRGBBCT-UHFFFAOYSA-N
SMILESOC(=O)CC1=CSC2=CC=C(Cl)C=C12
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Melting Point (clear melt)151.5-157.5?C
Appearance (Color)White to cream to yellow
Assay (Aqueous acid-base Titration)≥96.0 to ≤104.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only