Thermo Scientific Chemicals

2-Amino-3,6-dichlorobenzoic acid, 95%, Thermo Scientific Chemicals

Catalog number: L12850.06
5 g, Each
Thermo Scientific Chemicals

2-Amino-3,6-dichlorobenzoic acid, 95%, Thermo Scientific Chemicals

Catalog number: L12850.06
5 g, Each
Quantity
Catalog number: L12850.06
also known as L12850-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3032-32-4
Assay (Aqueous acid-base Titration)
≥94.0 to ≤106.0%
Appearance (Color)
Pale cream to cream to yellow to brown
Form
Powder or granules
Assay (HPLC)
≥94.0%

Description

2-Amino-3,6-dichlorobenzoic acid is used as a pharmaceutical intermediate and also finds use in chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Amino-3,6-dichlorobenzoic acid is used as a pharmaceutical intermediate and also finds use in chemical research.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents, acids, bases and sodium hypochlorite.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text