Thiophene-2-carboxylic anhydride is used as an organic chemical synthesis intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Thiophene-2-carboxylic anhydride is used as an organic chemical synthesis intermediate.
Solubility
It hydrolyzes in water.
Notes
Moisture sensitive. Incompatible with oxidizing agents.
RUO – Research Use Only
Weiwei Jin; Zhengkun Yu; Wei He; Wenjing Ye; Wen-Jing Xiao. Efficient Rh(I)-catalyzed direct arylation and alkenylation of arene C-H bonds via decarbonylation of benzoic and cinnamic anhydrides. Org. Lett. 2009, 11,(6), 1317-1320.