Thermo Scientific Chemicals

3-Hydroxy-2,2-dimethylpropionic acid, 97+%, Thermo Scientific Chemicals

Catalog number: L12858.18
50 g, Each
Thermo Scientific Chemicals

3-Hydroxy-2,2-dimethylpropionic acid, 97+%, Thermo Scientific Chemicals

Catalog number: L12858.18
50 g, Each
Quantity
Catalog number: L12858.18
also known as L12858-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4835-90-9
IUPAC Name
3-hydroxy-2,2-dimethylpropanoic acid
Molecular Formula
C5H10O3
InChI Key
RDFQSFOGKVZWKF-UHFFFAOYSA-N
SMILES
CC(C)(CO)C(O)=O
Form
Crystals or powder or crystalline powder
Assay (Silylated GC)
≥97.5%
Water Content (Karl Fischer Titration)
≤1%
Assay (Aqueous acid-base Titration)
≥97.5 to ≤102.5%
Identification (FTIR)
Conforms

Description

3-Hydroxy-2,2-dimethylpropionic acid is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Hydroxy-2,2-dimethylpropionic acid is used as pharmaceutical intermediate.

Solubility
Soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text