Thermo Scientific Chemicals

2-Benzyloxybenzaldehyde, 98%, Thermo Scientific Chemicals

Catalog number: L12866.09
10 g, Each
Thermo Scientific Chemicals

2-Benzyloxybenzaldehyde, 98%, Thermo Scientific Chemicals

Catalog number: L12866.09
10 g, Each
Quantity
Catalog number: L12866.09
also known as L12866-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5896-17-3
IUPAC Name
2-(benzyloxy)benzaldehyde
Molecular Formula
C14H12O2
InChI Key
PBEJTRAJWCNHRS-UHFFFAOYSA-N
SMILES
O=CC1=CC=CC=C1OCC1=CC=CC=C1
Appearance (Color)
White to pale cream
Assay (GC)
≥97.5%
Melting Point
45-50°C
Form
Crystalline powder or powder
Free acid (titration)
≤1.0%

Description

2-Benzyloxybenzaldehyde is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Benzyloxybenzaldehyde is used as an organic chemical synthesis intermediate.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text