Thermo Scientific Chemicals

6-Bromo-1-hexene, 95%, Thermo Scientific Chemicals

Catalog number: L12909.06
5 g, Each
Thermo Scientific Chemicals

6-Bromo-1-hexene, 95%, Thermo Scientific Chemicals

Catalog number: L12909.06
5 g, Each
Quantity
Catalog number: L12909.06
also known as L12909-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2695-47-8
IUPAC Name
6-bromohex-1-ene
Molecular Formula
C6H11Br
InChI Key
RIMXEJYJXDBLIE-UHFFFAOYSA-N
SMILES
BrCCCCC=C
Appearance (Color)
Clear colorless to yellow
Comment
Specification differs for U.S. and non-U.S. material where indicated
Assay (GC)
≥94.0%
Refractive Index
1.4630-1.4690 @ 20°C (non-U.S. specification)
Assay (GC)
≥94.0%

Description

6-Bromo-1-hexene was used in the preparation of epoxide, 6-bromo-1,2-epoxyhexane. It was also used in a simple, direct preparation of 4-alkenylbenzonitriles via anionic reduced forms of terephthalonitrile in liquid ammonia.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Bromo-1-hexene was used in the preparation of epoxide, 6-bromo-1,2-epoxyhexane. It was also used in a simple, direct preparation of 4-alkenylbenzonitriles via anionic reduced forms of terephthalonitrile in liquid ammonia.

Solubility
Not miscible in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text