4-Nitrophenyl-beta-D-galactopyranoside hydrate, 98+%, Thermo Scientific Chemicals
4-Nitrophenyl-beta-D-galactopyranoside hydrate, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Nitrophenyl-beta-D-galactopyranoside hydrate, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L12911.03
also known as L12911-03
Price (USD)/ Each
43.30
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
43.30
Add to cart
4-Nitrophenyl-beta-D-galactopyranoside hydrate, 98+%, Thermo Scientific Chemicals
Catalog numberL12911.03
Price (USD)/ Each
43.30
-
Add to cart
Chemical Identifiers
CAS200422-18-0
IUPAC Name(2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-(4-nitrophenoxy)oxane-3,4,5-triol
Molecular FormulaC12H15NO8
InChI KeyIFBHRQDFSNCLOZ-MFZRPPIDNA-N
SMILESOC[C@H]1O[C@@H](OC2=CC=C(C=C2)[N+]([O-])=O)[C@H](O)[C@@H](O)[C@H]1O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)≥98.0%
Appearance (Color)White to pale cream
FormPowder
Water Content (Karl Fischer Titration)4.0-7.0%
Optical Rotation-84.5? ± 0.5? (c=1 in water)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only