Thermo Scientific Chemicals

Domiphen bromide, 97%, Thermo Scientific Chemicals

Catalog number: L12930.14
25 g, Each
Thermo Scientific Chemicals

Domiphen bromide, 97%, Thermo Scientific Chemicals

Catalog number: L12930.14
25 g, Each
Quantity
Catalog number: L12930.14
also known as L12930-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
538-71-6
IUPAC Name
dodecyldimethyl(2-phenoxyethyl)azanium bromide
Molecular Formula
C22H40BrNO
InChI Key
OJIYIVCMRYCWSE-UHFFFAOYSA-M
SMILES
[Br-].CCCCCCCCCCCC[N+](C)(C)CCOC1=CC=CC=C1
Appearance (Color)
White to pale yellow
Form
Crystals or powder or crystalline powder or flakes
Assay (Titration ex Bromide)
≥96.0 to ≤104.0%
Loss on Drying
≤5.0%

Description

Domiphen bromide is employed as a phase transfer catalyst in synthetic chemistry. It is used as a quaternary ammonium cation with cationic surfactant properties. Further, it is used as an antiseptic and an active pharmaceutical ingredient.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Domiphen bromide is employed as a phase transfer catalyst in synthetic chemistry. It is used as a quaternary ammonium cation with cationic surfactant properties. Further, it is used as an antiseptic and an active pharmaceutical ingredient.

Solubility
Soluble in ethanol, acetone, ethyl acetate, chloroform and water. Slightly soluble in benzene.

Notes
Hygroscopic. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text