Thermo Scientific Chemicals

4-Phenyl-1-butyne, 98%, Thermo Scientific Chemicals

Catalog number: L12946.14
25 g, Each
Thermo Scientific Chemicals

4-Phenyl-1-butyne, 98%, Thermo Scientific Chemicals

Catalog number: L12946.14
25 g, Each
Quantity
Catalog number: L12946.14
also known as L12946-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
16520-62-0
IUPAC Name
(but-3-yn-1-yl)benzene
Molecular Formula
C10H10
InChI Key
QDEOKXOYHYUKMS-UHFFFAOYSA-N
SMILES
C#CCCC1=CC=CC=C1
Identification (FTIR)
Conforms
Refractive Index
1.5200-1.5240 @ 20°C
Appearance (Color)
Clear colorless to yellow
Form
Liquid
Assay (GC)
≥96.0%

Description

4-Phenyl-1-butyne is used as an intermediate in chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Phenyl-1-butyne is used as an intermediate in chemical research.

Solubility
Not miscible or difficult to mix in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text