Thermo Scientific Chemicals

(1S,2S)-(-)-1,2-Diphenyl-1,2-ethanediamine, 97%, Thermo Scientific Chemicals

Catalog number: L12968.14
25 g, Each
Thermo Scientific Chemicals

(1S,2S)-(-)-1,2-Diphenyl-1,2-ethanediamine, 97%, Thermo Scientific Chemicals

Catalog number: L12968.14
25 g, Each
Quantity
Catalog number: L12968.14
also known as L12968-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
29841-69-8
IUPAC Name
(1S,2S)-1,2-diphenylethane-1,2-bis(aminium)
Molecular Formula
C14H18N2
InChI Key
PONXTPCRRASWKW-KBPBESRZSA-P
SMILES
[NH3+][C@H]([C@@H]([NH3+])C1=CC=CC=C1)C1=CC=CC=C1
Assay (Non-aqueous acid-base Titration)
≥96.0 to ≤104.0%
Appearance (Color)
White to pale cream to cream to yellow
Form
Crystals or powder or crystalline powder or lumps or fused solid
Assay (GC)
≥96.0%
Melting Point (clear melt)
77.0-86.0?C

Description

(1S,2S)-(-)-1,2-Diphenyl-1,2-ethanediamine solvation agent, For synthesis of enantiopure ethylenediamines by chirality transfer (condensation with diketones followed by reductive cleavage), Co-catalyst in the Ru catalyzed enantioselective hydrogenation of aromatic ketones, Versatile ligand for the formation of metal complexes.1 Used in the synthesis of chiral tropocoronands which have potential utility in asymmetric catalysis

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(1S,2S)-(-)-1,2-Diphenyl-1,2-ethanediamine solvation agent, For synthesis of enantiopure ethylenediamines by chirality transfer (condensation with diketones followed by reductive cleavage), Co-catalyst in the Ru catalyzed enantioselective hydrogenation of aromatic ketones, Versatile ligand for the formation of metal complexes.1 Used in the synthesis of chiral tropocoronands which have potential utility in asymmetric catalysis

Solubility
Insoluble in water.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers. It is sensitive to air.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text