Thermo Scientific Chemicals

cis-1,4-Diacetoxy-2-butene, 96%, Thermo Scientific Chemicals

Catalog number: L13024.18
50 g, Each
Thermo Scientific Chemicals

cis-1,4-Diacetoxy-2-butene, 96%, Thermo Scientific Chemicals

Catalog number: L13024.18
50 g, Each
Quantity
Catalog number: L13024.18
also known as L13024-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
25260-60-0
IUPAC Name
(2Z)-4-(acetyloxy)but-2-en-1-yl acetate
Molecular Formula
C8H12O4
InChI Key
VZUAUHWZIKOMFC-ARJAWSKDSA-N
SMILES
CC(=O)OC\C=C/COC(C)=O
Appearance (Color)
Clear colorless to yellow
Assay (GC)
≥94.0%
Refractive Index
1.4390-1.4465 @ 20°C
Form
Liquid

Description

cis-1,4-Diacetoxy-2-butene is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
cis-1,4-Diacetoxy-2-butene is used as an organic chemical synthesis intermediate.

Solubility
Not miscible in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text