Thermo Scientific Chemicals

3-Penten-2-one, tech. 85%, Thermo Scientific Chemicals

Catalog number: L13031.03
1 g, Each
Thermo Scientific Chemicals

3-Penten-2-one, tech. 85%, Thermo Scientific Chemicals

Catalog number: L13031.03
1 g, Each
Quantity
Catalog number: L13031.03
also known as L13031-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
625-33-2
IUPAC Name
(3Z)-pent-3-en-2-one
Molecular Formula
C5H8O
InChI Key
LABTWGUMFABVFG-ARJAWSKDSA-N
SMILES
CC=C/C(C)=O
Assay (GC)
>83.0%
Form
liquid
Appearance (Color)
Clear, colourless to yellow
Refractive Index
1.4305 - 1.4405

Description

3-Penten-2-one

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Penten-2-one

Solubility
Slightly soluble in Water

Notes
Store at room temperature. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Incompatible with bases, reducing agents and oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text