Thermo Scientific Chemicals

Sodium ethoxide, 21% w/w in ethanol, Thermo Scientific Chemicals

Catalog number: L13083.AP
500 mL, Each
Thermo Scientific Chemicals

Sodium ethoxide, 21% w/w in ethanol, Thermo Scientific Chemicals

Catalog number: L13083.AP
500 mL, Each
Quantity
Catalog number: L13083.AP
also known as L13083-AP
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
141-52-6
IUPAC Name
sodium ethanolate
Molecular Formula
C2H5NaO
InChI Key
QDRKDTQENPPHOJ-UHFFFAOYSA-N
SMILES
[Na+].CC[O-]
Appearance (Color)
Clear pale yellow to orange to brown
Form
Liquid
Assay (Non-aqueous acid-base Titration)
≥20.0 to ≤22.0% (Sodium ethoxide)
Impurity content
≤0.50% [NaOH + Na2CO3 (as NaOH)]

Description

Sodium ethoxide, 21% w/w in ethanol is used as a strong base in organic synthesis. It finds application in various chemical reactions such as condensation, esterification, alkoxylation and etherifcation. It is actively involved in Claisen condensation, Stobbe reaction and Wolf-kishner reduction. It is an important starting material for the synthesis of ethyl ester and diethyl ester of malonic acid. In Williamson ether synthesis, it reacts with ethyl bromide to form diethyl ether.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium ethoxide, 21% w/w in ethanol is used as a strong base in organic synthesis. It finds application in various chemical reactions such as condensation, esterification, alkoxylation and etherifcation. It is actively involved in Claisen condensation, Stobbe reaction and Wolf-kishner reduction. It is an important starting material for the synthesis of ethyl ester and diethyl ester of malonic acid. In Williamson ether synthesis, it reacts with ethyl bromide to form diethyl ether.

Solubility
Miscible with alcohol.

Notes
Moisture sensitive. It hydrolyzes rapidly into sodium hydroxide in the presence of moist air. Incompatible with water, strong bases, acids, oxidizing agents, alkali metals, strong oxidizing agents, ammonia, acid chlorides, acid anhydrides, reducing agents, peroxides, acids and chlorinated solvents.
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
For Manufacturing and Laboratory Use. Not for beverage use

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text