Thermo Scientific Chemicals

Ethyl mesitylglyoxylate, 97%, Thermo Scientific Chemicals

Catalog number: L13106.06
5 g, Each
Thermo Scientific Chemicals

Ethyl mesitylglyoxylate, 97%, Thermo Scientific Chemicals

Catalog number: L13106.06
5 g, Each
Quantity
Catalog number: L13106.06
also known as L13106-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5524-57-2
IUPAC Name
ethyl 2-oxo-2-(2,4,6-trimethylphenyl)acetate
Molecular Formula
C13H16O3
InChI Key
XRSPPFOBPRPYLD-UHFFFAOYSA-N
SMILES
CCOC(=O)C(=O)C1=C(C)C=C(C)C=C1C
Assay (GC)
≥96.0%
Form
Liquid
Appearance (Color)
Clear pale yellow to yellow
Refractive Index
1.5100-1.5150 @ 20?C

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text