Thermo Scientific Chemicals

5-(2-Thienyl)pentanoic acid, 95%, Thermo Scientific Chemicals

Catalog number: L13122.06
5 g, Each
Thermo Scientific Chemicals

5-(2-Thienyl)pentanoic acid, 95%, Thermo Scientific Chemicals

Catalog number: L13122.06
5 g, Each
Quantity
Catalog number: L13122.06
also known as L13122-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
21010-06-0
IUPAC Name
5-(thiophen-2-yl)pentanoic acid
Molecular Formula
C9H12O2S
InChI Key
OQFTZRHAQGXEQU-UHFFFAOYSA-N
SMILES
OC(=O)CCCCC1=CC=CS1
Assay (Aqueous acid-base Titration)
≥94.0 to ≤106.0%
Appearance (Color)
Pale cream to yellow to brown
Form
Crystals or powder or crystalline powder
Assay (GC)
≥94.0%
Melting Point (clear melt)
34.0-46.0?C

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text