Thermo Scientific Chemicals

2-Amino-4,4,4-trifluorobutyric acid, 97%, Thermo Scientific Chemicals

Catalog number: L13131.MC
100 mg, Each
Thermo Scientific Chemicals

2-Amino-4,4,4-trifluorobutyric acid, 97%, Thermo Scientific Chemicals

Catalog number: L13131.MC
100 mg, Each
Quantity
Catalog number: L13131.MC
also known as L13131-MC
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
15959-93-0
IUPAC Name
2-amino-4,4,4-trifluorobutanoic acid
Molecular Formula
C4H6F3NO2
InChI Key
AQPCXCOPDSEKQT-UHFFFAOYNA-N
SMILES
NC(CC(F)(F)F)C(O)=O
Assay (GC)
≥96.0%
Appearance (Color)
White to cream to pale yellow
Form
Crystals or powder or crystalline powder
Identification (FTIR)
Conforms
Assay (Aqueous acid-base Titration)
≥96.0 to ≤104.0%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text