Thermo Scientific Chemicals

2(5H)-Furanone, 96%, Thermo Scientific Chemicals

Catalog number: L13142.06
5 g, Each
Thermo Scientific Chemicals

2(5H)-Furanone, 96%, Thermo Scientific Chemicals

Catalog number: L13142.06
5 g, Each
Quantity
Catalog number: L13142.06
also known as L13142-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
497-23-4
IUPAC Name
2,5-dihydrofuran-2-one
Molecular Formula
C4H4O2
InChI Key
VIHAEDVKXSOUAT-UHFFFAOYSA-N
SMILES
O=C1OCC=C1
Appearance (Color)
Clear colorless to yellow
Form
Liquid
Assay (GC)
>95.0%
Refractive Index
1.4670-1.4710 @ 20?C

Description

2(5H)-Furanone is employed in the preparation of 5-substituted 2(5H) furanones (gamma-butenolides) catalyzed by bifunctional aminothiourea and aminosquaramide organocatalysts through direct aldol reaction with aromatic aldehydes. Further, it is used in the synthesis of lignans by Michael addition reactions. In addition to this, it is used in three-component Michael-Aldol reactions with an aldehyde and thiolate or carbanion.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2(5H)-Furanone is employed in the preparation of 5-substituted 2(5H) furanones (gamma-butenolides) catalyzed by bifunctional aminothiourea and aminosquaramide organocatalysts through direct aldol reaction with aromatic aldehydes. Further, it is used in the synthesis of lignans by Michael addition reactions. In addition to this, it is used in three-component Michael-Aldol reactions with an aldehyde and thiolate or carbanion.

Solubility
Immiscible with water.

Notes
Light and moisture sensitive. Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text