Thermo Scientific Chemicals

3-(3-Furyl)acrylic acid, 98%, Thermo Scientific Chemicals

Catalog number: L13143.14
25 g, Each
Thermo Scientific Chemicals

3-(3-Furyl)acrylic acid, 98%, Thermo Scientific Chemicals

Catalog number: L13143.14
25 g, Each
Quantity
Catalog number: L13143.14
also known as L13143-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
39244-10-5
Appearance (Color)
White to cream to brown
Form
Crystals or powder or crystalline powder or granules
Assay (Aqueous acid-base Titration)
≥97.5 to ≤102.5%
Assay (Silylated GC)
≥97.5%
Melting Point (clear melt)
148-156°C

Description

3-(3-Furyl)acrylic acid is used as an intermediate in chemical research and pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-(3-Furyl)acrylic acid is used as an intermediate in chemical research and pharmaceuticals.

Solubility
Sparingly soluble in water (0.56 g/L at 25°C).

Notes
Store at room temperature. Keep container tightly closed. Store in cool, dry conditions in well sealed containers. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text