Thermo Scientific Chemicals

6-Heptenoic acid, 96%, Thermo Scientific Chemicals

Catalog number: L13144.06
5 g, Each
Thermo Scientific Chemicals

6-Heptenoic acid, 96%, Thermo Scientific Chemicals

Catalog number: L13144.06
5 g, Each
Quantity
Catalog number: L13144.06
also known as L13144-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1119-60-4
IUPAC Name
hept-6-enoic acid
Molecular Formula
C7H12O2
InChI Key
RWNJOXUVHRXHSD-UHFFFAOYSA-N
SMILES
OC(=O)CCCCC=C
Form
Liquid
Assay (Aqueous acid-base Titration)
≥95.0 to ≤105.0%
Assay (GC)
≥95.0%
Appearance (Color)
Clear colorless
Identification (FTIR)
Conforms

Description

6-Heptenoic acid is used in the preparation of 6-(iodomethyl)-hexanolide through iodo-lactonization.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Heptenoic acid is used in the preparation of 6-(iodomethyl)-hexanolide through iodo-lactonization.

Solubility
Miscible with water.

Notes
Incompatible with strong oxidizing agents, strong acids and bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text