Thermo Scientific Chemicals

L-Leucine ethyl ester hydrochloride, 97%, Thermo Scientific Chemicals

Catalog number: L13147.06
5 g, Each
Thermo Scientific Chemicals

L-Leucine ethyl ester hydrochloride, 97%, Thermo Scientific Chemicals

Catalog number: L13147.06
5 g, Each
Quantity
Catalog number: L13147.06
also known as L13147-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2743-40-0
IUPAC Name
ethyl (2S)-2-amino-4-methylpentanoate
Molecular Formula
C8H17NO2
InChI Key
QIGLJVBIRIXQRN-ZETCQYMHSA-N
SMILES
CCOC(=O)[C@@H](N)CC(C)C
Appearance (Color)
White
Form
Crystals or powder or crystalline powder or fused solid or needles
Assay (Titration ex Chloride)
≥96.0 to ≤104.0%
Melting Point (clear melt)
134-138°C
Optical Rotation
+16.5° to +19.5° (c=5 in ethanol)

Description

L-Leucine ethyl ester hydrochloride is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Leucine ethyl ester hydrochloride is used as an organic chemical synthesis intermediate.

Solubility
Soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text