Thermo Scientific Chemicals

1-Phenyl-1,4-pentanedione, 96%, Thermo Scientific Chemicals

Catalog number: L13151.03
1 g, Each
Thermo Scientific Chemicals

1-Phenyl-1,4-pentanedione, 96%, Thermo Scientific Chemicals

Catalog number: L13151.03
1 g, Each
Quantity
Catalog number: L13151.03
also known as L13151-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
583-05-1
IUPAC Name
1-phenylpentane-1,4-dione
Molecular Formula
C11H12O2
InChI Key
RBLXWIPBPPVLPU-UHFFFAOYSA-N
SMILES
CC(=O)CCC(=O)C1=CC=CC=C1
Form
Liquid
Assay (GC)
≥95.0%
Appearance (Color)
Clear colourless to yellow or pale orange
Refractive Index
1.5300-1.5360 @20°C

Description

1-Phenyl-1,4-pentanedione is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Phenyl-1,4-pentanedione is used as an organic chemical synthesis intermediate.

Solubility
Not miscible in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text