Thermo Scientific Chemicals

N-(Benzyloxycarbonyl)hydroxylamine, 98+%, Thermo Scientific Chemicals

Catalog number: L13175.14
25 g, Each
Thermo Scientific Chemicals

N-(Benzyloxycarbonyl)hydroxylamine, 98+%, Thermo Scientific Chemicals

Catalog number: L13175.14
25 g, Each
Quantity
Catalog number: L13175.14
also known as L13175-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3426-71-9
IUPAC Name
benzyl N-hydroxycarbamate
Molecular Formula
C8H9NO3
InChI Key
PQBSPTAPCMSZAA-UHFFFAOYSA-N
SMILES
ONC(=O)OCC1=CC=CC=C1
Melting Point (clear melt)
64.0-70.0?C
Appearance (Color)
White to cream
Assay (HPLC)
≥98.0%
Form
Crystals or powder or crystalline powder

Description

N-(Benzyloxycarbonyl)hydroxylamine reacts with cyclohepta-1,3-diene to produce 6-oxa-7-aza-bicyclo[3.2.2]non-8-ene-7-carboxylic acid benzyl ester.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-(Benzyloxycarbonyl)hydroxylamine reacts with cyclohepta-1,3-diene to produce 6-oxa-7-aza-bicyclo[3.2.2]non-8-ene-7-carboxylic acid benzyl ester.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. It is sensitive to moisture. Incompatible with strong oxidizing agents. Store at -20°C.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text