Thermo Scientific™

Borane-pyridine complex, 95%, Thermo Scientific Chemicals

Catalog number: L13178.09
10 g, Each
Thermo Scientific™

Borane-pyridine complex, 95%, Thermo Scientific Chemicals

Catalog number: L13178.09
10 g, Each
Quantity
Catalog number: L13178.09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
110-51-0
IUPAC Name
(pyridin-1-ium-1-yl)boranuide
Molecular Formula
C5H8BN
InChI Key
LPGWNCNRGQANGC-UHFFFAOYSA-N
SMILES
[BH3-][N+]1=CC=CC=C1
Appearance (Color)
Clear, colourless to yellow
Assay (unspecified)
> 94.0%
Form
liquid

Description

Borane-pyridine complex is used for synthesis of α-alkyl-β-hydroxy esters and preparation of Igs conjugated with viral peptide epitopes. It is used as reducing agent for high performance liquid chromatography analysis of oligosaccharides. It is also used as modifier in decant oil affecting its carbonization to mesophase pitch.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Borane-pyridine complex is used for synthesis of α-alkyl-β-hydroxy esters and preparation of Igs conjugated with viral peptide epitopes. It is used as reducing agent for high performance liquid chromatography analysis of oligosaccharides. It is also used as modifier in decant oil affecting its carbonization to mesophase pitch.

Solubility
It hydrolyzes slowly in water.

Notes
Air and moisture sensitive. Incompatible with oxidizing agents and heat.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text