Butyric acid, 99+%, Thermo Scientific Chemicals
Butyric acid, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Butyric acid, 99+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2500 mL
100 mL
500 mL
Catalog number L13189.0F
Price (USD)/ Each
69.65
Online exclusive
76.80 
Save 7.15 (9%)
-
Add to cart
Quantity:
2500 mL
Request bulk or custom format
Price (USD)/ Each
69.65
Online exclusive
76.80 
Save 7.15 (9%)
Add to cart
Butyric acid, 99+%, Thermo Scientific Chemicals
Catalog numberL13189.0F
Price (USD)/ Each
69.65
Online exclusive
76.80 
Save 7.15 (9%)
-
Add to cart
Chemical Identifiers
CAS107-92-6
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.3965-1.3990 @ 20?C
Assay (GC)≥99.0%
Appearance (Color)Clear colorless
Assay (Aqueous acid-base Titration)≥99.0 to ≤101.0%
Identification (FTIR)Conforms
View more
Butyric acid is a useful as precursor for the preparation of various butyrate esters. It is used as a food and perfume additive. It is also used as an animal feed supplement. Due to its powerful odor, it has also been used as a fishing bait additive and as a stink bomb. It is widely used as a flavoring agent for human food and beverages.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Butyric acid is a useful as precursor for the preparation of various butyrate esters. It is used as a food and perfume additive. It is also used as an animal feed supplement. Due to its powerful odor, it has also been used as a fishing bait additive and as a stink bomb. It is widely used as a flavoring agent for human food and beverages.

Solubility
Miscible with water, ethanol and ether. Slightly miscible with carbon tetrachloride.

Notes
Store in cool place. Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Ching, S.; Richter, I. J.; Tutunjian, K. A.; Kriz, D. A.; Kovic, Y. Synthesis of highly monodisperse porous manganese oxide spheres using a butyric acid microemulsion. Chem. Commun. 2015, 51 (10), 1961-1964.
  2. Bianchi, N.; Chiarabelli, C.; Zuccato, C.; Lampronti, I.; Borgatti, M.; Amari, G.; Delcanale, M.; Chiavilli, F.; Prus, E.; Fibach, E.; Gambari, R. Erythroid differentiation ability of butyric acid analogues: Identification of basal chemical structures of new inducers of foetal haemoglobin. Mol. Cell. Pharm. 2015, 752, 84-91.