Fluorescein
Thermo Scientific Chemicals

Fluorescein

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number L13251.22
also known as L13251-22
Price (USD)/ Each
39.65
Online exclusive
43.60 
Save 3.95 (9%)
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
39.65
Online exclusive
43.60 
Save 3.95 (9%)
Add to cart
Fluorescein
Catalog numberL13251.22
Price (USD)/ Each
39.65
Online exclusive
43.60 
Save 3.95 (9%)
-
Add to cart
Chemical Identifiers
CAS2321-07-5
IUPAC Name3',6'-dihydroxy-3H-spiro[2-benzofuran-1,9'-xanthen]-3-one
Molecular FormulaC20H12O5
InChI KeyGNBHRKFJIUUOQI-UHFFFAOYSA-N
SMILESOC1=CC=C2C(OC3=CC(O)=CC=C3C22OC(=O)C3=CC=CC=C23)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Loss on Drying≤3%
lambda-max (UV-Vis)490-492 nm (0.1N NaOH)
Specific Absorbance (E 1%/1cm)≥1800 @ Lambda max (0.1N NaOH)
Appearance (Color)Orange to red to brown
Fluorescein is used as a fluorescent tracer for many applications including in a type of dye laser as the gain medium, in forensics and serology to detect latent blood stains and in dye tracing. It is used to localise multiple muscular ventricular septal defects during open heart surgery and confirm the presence of any residual defects. It is applied to teeth to reveal plaque.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Slightly soluble in water and alcohol.

Notes
Incompatibles with strong oxidizing agents. Moisture sensitive.
RUO – Research Use Only

General References:

  1. Akram, A. C.; Noman, S.; Moniri-Javid, R.; Gizicki, J. P.; Reed, E. A.; Singh, S. B.; Basu, A. S.; Banno, F.; Fujimoto, M.; Ram, J. L. Development of an automated ballast water treatment verification system utilizing fluorescein diacetate hydrolysis as a measure of treatment efficacy. Water Res. 2015, 70, 404-413.
  2. Tichauer, K. M.; Guthrie, M.; Hones, L.; Sinha, L.; St. Lawrence, K.; Kang-Mieler, J. J. Quantitative retinal blood flow mapping from fluorescein videoangiography using tracer kinetic modeling. Opt. Lett. 2015, 40 (10), 2169-2172.