Thermo Scientific Chemicals

Fluorescein

Catalog number: L13251.36
500 g, Each
Thermo Scientific Chemicals

Fluorescein

Catalog number: L13251.36
500 g, Each
Quantity
Catalog number: L13251.36
also known as L13251-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2321-07-5
IUPAC Name
3',6'-dihydroxy-3H-spiro[2-benzofuran-1,9'-xanthen]-3-one
Molecular Formula
C20H12O5
InChI Key
GNBHRKFJIUUOQI-UHFFFAOYSA-N
SMILES
OC1=CC=C2C(OC3=CC(O)=CC=C3C22OC(=O)C3=CC=CC=C23)=C1
Form
Powder
Loss on Drying
≤3%
lambda-max (UV-Vis)
490-492 nm (0.1N NaOH)
Specific Absorbance (E 1%/1cm)
≥1800 @ Lambda max (0.1N NaOH)
Appearance (Color)
Orange to red to brown

Description

Fluorescein is used as a fluorescent tracer for many applications including in a type of dye laser as the gain medium, in forensics and serology to detect latent blood stains and in dye tracing. It is used to localise multiple muscular ventricular septal defects during open heart surgery and confirm the presence of any residual defects. It is applied to teeth to reveal plaque.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Slightly soluble in water and alcohol.

Notes
Incompatibles with strong oxidizing agents. Moisture sensitive.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text