Thermo Scientific Chemicals

Methanol, 99%, Thermo Scientific Chemicals

Catalog number: L13255.AP
500 mL, Each
Thermo Scientific Chemicals

Methanol, 99%, Thermo Scientific Chemicals

Catalog number: L13255.AP
500 mL, Each
Quantity
Catalog number: L13255.AP
also known as L13255-AP
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
67-56-1
IUPAC Name
methanol
Molecular Formula
CH4O
InChI Key
OKKJLVBELUTLKV-UHFFFAOYSA-N
SMILES
CO
Comment
Specification differs for U.S. and non-U.S. material where indicated
Identification (FTIR)
Conforms (non-U.S. specification)
Refractive Index
1.3275-1.3305 @ 20°C (non-U.S. specification)
Appearance (Color)
Clear, colorless
Assay (GC)
≥98.5% (non-U.S. specification)

Description

Methanol is useful in the synthesis of nucleic acids and peptides. It is used as an antifreeze, solvent, fuel, and as a denaturant for ethanol. It is also used for producing biodiesel. It is used for the synthesis of formaldehyde and acetic acid. It is also useful to make chemicals, to remove water from automotive and aviation fuels, as a solvent for paints and plastics, and as an ingredient in a wide variety of products. It plays an important role in the measurement of methyl ester content of pectin. Further, it is widely used as a solvent in synthetic organic chemistry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methanol is useful in the synthesis of nucleic acids and peptides. It is used as an antifreeze, solvent, fuel, and as a denaturant for ethanol. It is also used for producing biodiesel. It is used for the synthesis of formaldehyde and acetic acid. It is also useful to make chemicals, to remove water from automotive and aviation fuels, as a solvent for paints and plastics, and as an ingredient in a wide variety of products. It plays an important role in the measurement of methyl ester content of pectin. Further, it is widely used as a solvent in synthetic organic chemistry.

Solubility
Miscible with water and most of the organic solvents including ethyl acetate, toluene, dichloromethane, chloroform, tetrahydrofuran, and acetone.

Notes
Hygroscopic. Keep the material away from heat, sparks, flame and other sources of ignition.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text