2-Bromo-4'-(diethylamino)acetophenone, is used as non-ionic dialyzable detergent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Bromo-4′-(diethylamino)acetophenone, is used as non-ionic dialyzable detergent.
Solubility
Insoluble in water.
Notes
Heat and moisture sensitive. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
RUO – Research Use Only
JM Holub.; K O’Toole-Colin.; A Getzel.; A Argenti. Lipid-lowering effects of ethyl 2-phenacyl-3-aryl-1H-pyrrole-4-carboxylates in rodents. Molecules. 2004, 9,(3), 134-157.
M Soural.;V Krchnák. Efficient Solid-Phase Synthesis of 2-Substituted-3-Hydroxy-4 (1 H)-Quinolinone-7-Carboxamides with Two Diversity Positions. Journal of combinatorial chemistry. 2007, 9,(5), 793-796.
Nonionic dialyzable detergent.