Thermo Scientific Chemicals

2-Pentanone, 99+%, Thermo Scientific Chemicals

Catalog number: L13262.14
25 g, Each
Thermo Scientific Chemicals

2-Pentanone, 99+%, Thermo Scientific Chemicals

Catalog number: L13262.14
25 g, Each
Quantity
Catalog number: L13262.14
also known as L13262-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
107-87-9
IUPAC Name
pentan-2-one
Molecular Formula
C5H10O
InChI Key
XNLICIUVMPYHGG-UHFFFAOYSA-N
SMILES
CCCC(C)=O
Appearance (Color)
Clear colorless to pale yellow
Form
Liquid
Identification (FTIR)
Conforms (non-U.S. sourced material)
Assay from Supplier's CofA
≥98.5% (U.S. sourced material)
Refractive Index
1.3890-1.3910 @ 20°C (non-U.S. sourced material)

Description

2-Pentanone is used as a solvent for lacquers and surface coatings. It acts as a flavoring agent. It is also used as a solvent for cleaning and degreasing purpose. Further, it serves as an intermediate, paint additive and coating additive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Pentanone is used as a solvent for lacquers and surface coatings. It acts as a flavoring agent. It is also used as a solvent for cleaning and degreasing purpose. Further, it serves as an intermediate, paint additive and coating additive.

Solubility
Miscible with water.

Notes
Incompatible with strong bases, oxidizing agents and reducing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text