Thermo Scientific Chemicals

(S)-(+)-2-Phenylglycinol, 98+%, Thermo Scientific Chemicals

Catalog number: L13265.14
25 g, Each
Thermo Scientific Chemicals

(S)-(+)-2-Phenylglycinol, 98+%, Thermo Scientific Chemicals

Catalog number: L13265.14
25 g, Each
Quantity
Catalog number: L13265.14
also known as L13265-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
20989-17-7
IUPAC Name
(2S)-2-amino-2-phenylethan-1-ol
Molecular Formula
C8H11NO
InChI Key
IJXJGQCXFSSHNL-MRVPVSSYSA-N
SMILES
N[C@H](CO)C1=CC=CC=C1
Form
Powder or crystalline powder
Melting Point (clear melt)
74-80°C
Solution Test
Clear (2.5% in 1N HCl)
Appearance (Color)
White to pale yellow
Assay (Non-aqueous acid-base Titration)
≥98.0 to ≤102.0%

Description

(S)-(+)-2-Phenylglycinol is used in the synthesis of chiral, unsymmetrical bisoxazolines. It is used as a reagent for the resolution of acids via the easily hydrolyzed amides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(S)-(+)-2-Phenylglycinol is used in the synthesis of chiral, unsymmetrical bisoxazolines. It is used as a reagent for the resolution of acids via the easily hydrolyzed amides.

Solubility
Soluble in chloroform, dimethylsulfoxide and methanol. Limited solubility in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. It is sensitive to air. Incompatible with oxidizing agents. Store at -20°C.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text