Thermo Scientific Chemicals

Tetra-n-butylammonium fluoride trihydrate, 98%, Thermo Scientific Chemicals

Catalog number: L13303.18
50 g, Each
Thermo Scientific Chemicals

Tetra-n-butylammonium fluoride trihydrate, 98%, Thermo Scientific Chemicals

Catalog number: L13303.18
50 g, Each
Quantity
Catalog number: L13303.18
also known as L13303-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
87749-50-6
IUPAC Name
tetrabutylazanium trihydrate fluoride
Molecular Formula
C16H42FNO3
InChI Key
VEPTXBCIDSFGBF-UHFFFAOYSA-M
SMILES
O.O.O.[F-].CCCC[N+](CCCC)(CCCC)CCCC
Form
Crystals or crystalline powder and/or lumps or fused solid
Melting Point (clear melt)
49-60?C
Appearance (Color)
White to yellow
Assay (Non-aqueous acid-base Titration)
≥ 97.5 to ≤102.5% (as trihydrate)
Identification (FTIR)
Conforms

Description

Tetra-n-butylammonium fluoride trihydrate is used as a phase transfer catalyst, as a mild base and as a source of fluoride ion in organic solvents. It is also used as a deprotecting agent to remove silyl ether protecting groups. It acts as a reactant for the preparation of deprotecting agents, in preparation of cellulose derivatives, lipophilic peptides for DNA transfections and Dehydrobromination reactions. It plays an important role in the convertion of O-silylated enolates in to carbonyls in the presence of dimethyl sulfoxide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetra-n-butylammonium fluoride trihydrate is used as a phase transfer catalyst, as a mild base and as a source of fluoride ion in organic solvents. It is also used as a deprotecting agent to remove silyl ether protecting groups. It acts as a reactant for the preparation of deprotecting agents, in preparation of cellulose derivatives, lipophilic peptides for DNA transfections and Dehydrobromination reactions. It plays an important role in the convertion of O-silylated enolates in to carbonyls in the presence of dimethyl sulfoxide.

Solubility
Soluble in water, terahydrofuran, dimethyl sulfoxide and acetonitrile.

Notes
Hygroscopic. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text