Thermo Scientific Chemicals

Polysorbate 80, Thermo Scientific Chemicals

Catalog number: L13315.0F
2500 mL, Each
Thermo Scientific Chemicals

Polysorbate 80, Thermo Scientific Chemicals

Catalog number: L13315.0F
2500 mL, Each
Quantity
Catalog number: L13315.0F
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
9005-65-6
Molecular Formula
(C2H4O)x(C2H4O)z(C2H4O)y(C2H4O)wC24H44O6
InChI Key
HDTIFOGXOGLRCB-KTKRTIGZNA-N
SMILES
CCCCCCCC\C=C/CCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO
MDL Number
MFCD00082107
Appearance (Color)
Yellow to orange
Comment
Material sourced in the U.S. and in other countries
Refractive Index
1.4700-1.4760 @ 20°C (non-U.S. sourced material)
Form
Viscous liquid
Water Content (Karl Fischer Titration)
≤4.0%

Description

Used for solubilizing membrane proteins during isolation and purification.Polysorbate 80 is used as a surfactant in soaps, cosmetics, body washes, hair shampoos, hair loss treatments, lotions, creams, bath oils and in mouthwash in the health care industry. It serves as an emulsifier in foods such as ice cream and in the preparation of antiarrhythmic amiodarone. Further, it acts as a solubilizing agent and excipient in the pharmaceutical industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text