Thermo Scientific Chemicals

4-Vinylpyridine, 95%, stab. with 80 to 150ppm hydroquinone, Thermo Scientific Chemicals

Catalog number: L13316.AP
500 mL, Each
Thermo Scientific Chemicals

4-Vinylpyridine, 95%, stab. with 80 to 150ppm hydroquinone, Thermo Scientific Chemicals

Catalog number: L13316.AP
500 mL, Each
Quantity
Catalog number: L13316.AP
also known as L13316-AP
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
100-43-6
IUPAC Name
4-ethenylpyridine
Molecular Formula
C7H7N
InChI Key
KFDVPJUYSDEJTH-UHFFFAOYSA-N
SMILES
C=CC1=CC=NC=C1
Comment
May contain polymerised material and darken on storage
Stabilizer
Hydroquinone (80-150 ppm)
Appearance (Color)
Clear colorless to yellow to red/brown
Assay (GC)
≥94.0%
Identification (FTIR)
Conforms

Description

4-Vinylpyridine acts as a co-monomer in styrene-butadiene polymers to promote adhesion between the rubber compound and supporting fibers (or cords) in tires, belts and hoses. It serves as an intermediate for manufacturing chemicals and pharmaceuticals. Further, it is used as a reagent for the modification of cysteine. In addition to this, it is used to make poly(4-vinylpyridine) and elastomers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Vinylpyridine acts as a co-monomer in styrene-butadiene polymers to promote adhesion between the rubber compound and supporting fibers (or cords) in tires, belts and hoses. It serves as an intermediate for manufacturing chemicals and pharmaceuticals. Further, it is used as a reagent for the modification of cysteine. In addition to this, it is used to make poly(4-vinylpyridine) and elastomers.

Solubility
Miscible with water, chloroform, ethanol, dilute acids, alcohol, hydrocarbons, esters and ketones.

Notes
Light sensitive. Incompatible with strong acids, strong oxidizing agents, ultraviolet radiation and free radical initiators.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text