Allyl vinyl ether is used as a intermediate in chemical research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Allyl vinyl ether is used as a intermediate in chemical research.
Solubility
Slightly soluble in water.
Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents, ultraviolet radiation and free radical initiators.
RUO – Research Use Only
Daniel L. Severance.; William L. Jorgensen. Effects of hydration on the Claisen rearrangement of allyl vinyl ether from computer simulations. J. Am. Chem. Soc. 1986, 27 (51), 6267-6270.
J.L van der Baan.; F Bickelhaupt. Palladium(II)-catalyzed claisen rearrangement of allyl vinyl ethers. Tetrahedron Letters. 2015, 27 (51), 3458-3462.