Thermo Scientific Chemicals

2-Bromo-4'-(diethylamino)acetophenone, 98%, Thermo Scientific Chemicals

Catalog number: L13337.03
1 g, Each
Thermo Scientific Chemicals

2-Bromo-4'-(diethylamino)acetophenone, 98%, Thermo Scientific Chemicals

Catalog number: L13337.03
1 g, Each
Quantity
Catalog number: L13337.03
also known as L13337-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
207986-25-2
IUPAC Name
2-bromo-1-[4-(diethylamino)phenyl]ethan-1-one
Molecular Formula
C12H16BrNO
InChI Key
ALTMCDJIFXTEPV-UHFFFAOYSA-N
SMILES
CCN(CC)C1=CC=C(C=C1)C(=O)CBr
Appearance (Color)
Yellow to green
Form
Crystals or powder or crystalline powder or fused solid
Assay (HPLC)
≥97.5%

Description

2-Bromo-4'-(diethylamino)acetophenone, is used as a pharmaceutical intermediate. It is also used as a primary and secondary intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromo-4′-(diethylamino)acetophenone, is used as a pharmaceutical intermediate. It is also used as a primary and secondary intermediate.

Solubility
It is insoluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text