3,3',4,4'-Tetramethylbiphenyl, 98%, Thermo Scientific Chemicals
3,3',4,4'-Tetramethylbiphenyl, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3,3',4,4'-Tetramethylbiphenyl, 98%, Thermo Scientific Chemicals

 
Have Questions?
Quantity:
1 g
Catalog number L13338.03
also known as L13338-03
Price (USD)/ Each
53.65
Online exclusive
59.80 
Save 6.15 (10%)
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
53.65
Online exclusive
59.80 
Save 6.15 (10%)
Add to cart
3,3',4,4'-Tetramethylbiphenyl, 98%, Thermo Scientific Chemicals
Catalog numberL13338.03
Price (USD)/ Each
53.65
Online exclusive
59.80 
Save 6.15 (10%)
-
Add to cart
Chemical Identifiers
CAS4920-95-0
IUPAC Name3,3',4,4'-tetramethyl-1,1'-biphenyl
Molecular FormulaC16H18
InChI KeyYXBIAYXZUDJVEB-UHFFFAOYSA-N
SMILESCC1=CC=C(C=C1C)C1=CC=C(C)C(C)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
FormCrystals or powder or crystalline powder
Assay (GC)≥97.5%
Melting Point (clear melt)73.0-79.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only