Thermo Scientific Chemicals

4-Iodophenylacetic acid, 97%, Thermo Scientific Chemicals

Catalog number: L13345.06
5 g, Each
Thermo Scientific Chemicals

4-Iodophenylacetic acid, 97%, Thermo Scientific Chemicals

Catalog number: L13345.06
5 g, Each
Quantity
Catalog number: L13345.06
also known as L13345-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1798-06-7
IUPAC Name
2-(4-iodophenyl)acetic acid
Molecular Formula
C8H7IO2
InChI Key
FJSHTWVDFAUNCO-UHFFFAOYSA-N
SMILES
OC(=O)CC1=CC=C(I)C=C1
Appearance (Color)
White to cream to pale brown
Form
Powder or crystalline powder
Assay (Aqueous acid-base Titration)
≥96.0 to ≤104.0%
Assay (HPLC)
≥96.0%
Melting Point (clear melt)
136-143?C

Description

4-Iodophenylacetic acid, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also used in suzuki coupling reaction transport.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Iodophenylacetic acid, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also used in suzuki coupling reaction transport.

Solubility
Insoluble in water.

Notes
Light Sensitive. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text