Thermo Scientific Chemicals

Tris(hydroxymethyl)nitromethane, 95%, Thermo Scientific Chemicals

Catalog number: L13349.36
500 g, Each
Thermo Scientific Chemicals

Tris(hydroxymethyl)nitromethane, 95%, Thermo Scientific Chemicals

Catalog number: L13349.36
500 g, Each
Quantity
Catalog number: L13349.36
also known as L13349-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
126-11-4
IUPAC Name
2-(hydroxymethyl)-2-nitropropane-1,3-diol
Molecular Formula
C4H9NO5
InChI Key
OLQJQHSAWMFDJE-UHFFFAOYSA-N
SMILES
OCC(CO)(CO)[N+]([O-])=O
Assay (Silylated GC)
≥94.0%
Identification (FTIR)
Conforms
Appearance (Color)
White to pale cream to cream to yellow to orange to pale brown
Form
Crystals or powder or crystalline powder or lumps or flakes or granules

Description

Tris(hydroxymethyl)nitromethane, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is used as a primary and secondary intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tris(hydroxymethyl)nitromethane, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is used as a primary and secondary intermediate.

Solubility
It is soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents, bases. Stable under recommended storage conditions.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text