Thermo Scientific Chemicals

1-Dodecyl isocyanate, 98%, Thermo Scientific Chemicals

Catalog number: L13380.06
5 g, Each
Thermo Scientific Chemicals

1-Dodecyl isocyanate, 98%, Thermo Scientific Chemicals

Catalog number: L13380.06
5 g, Each
Quantity
Catalog number: L13380.06
also known as L13380-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4202-38-4
IUPAC Name
1-isocyanatododecane
Molecular Formula
C13H25NO
InChI Key
YIDSTEJLDQMWBR-UHFFFAOYSA-N
SMILES
CCCCCCCCCCCCN=C=O
Assay (GC)
> 97.5%
Appearance (Color)
Colourless
Form
Clear to slightly turbid liquid
Comment
May form slight sediment on storage, filter before use.
Refractive Index
1.4385 - 1.4435 @20?C

Description

1-Dodecyl isocyanate, is incorporated in AZT analogue preparation through it use ine the synthesis of carbodiimides from phosphinimines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Dodecyl isocyanate, is incorporated in AZT analogue preparation through it use ine the synthesis of carbodiimides from phosphinimines.

Solubility
It hydrolyzes in water.

Notes
Moisture Sensitive. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text