Thermo Scientific Chemicals

N,N,N',N'-Tetramethylthiourea, 98%, Thermo Scientific Chemicals

Catalog number: L13392.06
5 g, Each
Thermo Scientific Chemicals

N,N,N',N'-Tetramethylthiourea, 98%, Thermo Scientific Chemicals

Catalog number: L13392.06
5 g, Each
Quantity
Catalog number: L13392.06
also known as L13392-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2782-91-4
IUPAC Name
1,1,3,3-tetramethylthiourea
Molecular Formula
C5H12N2S
InChI Key
MNOILHPDHOHILI-UHFFFAOYSA-N
SMILES
CN(C)C(=S)N(C)C
Appearance (Color)
White to yellow
Assay (GC)
≥97.5%
Form
Crystals or powder or crystalline powder or lumps or fused solid

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text