N,N,N',N'-Tetramethylthiourea, 98%, Thermo Scientific Chemicals
N,N,N',N'-Tetramethylthiourea, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N,N,N',N'-Tetramethylthiourea, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number L13392.14
also known as L13392-14
Price (USD)/ Each
76.60
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
76.60
Add to cart
N,N,N',N'-Tetramethylthiourea, 98%, Thermo Scientific Chemicals
Catalog numberL13392.14
Price (USD)/ Each
76.60
-
Add to cart
Chemical Identifiers
CAS2782-91-4
IUPAC Name1,1,3,3-tetramethylthiourea
Molecular FormulaC5H12N2S
InChI KeyMNOILHPDHOHILI-UHFFFAOYSA-N
SMILESCN(C)C(=S)N(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow
Assay (GC)≥97.5%
FormCrystals or powder or crystalline powder or lumps or fused solid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only