Thermo Scientific Chemicals

1-Pyrrolidinecarbonyl chloride, 97%, Thermo Scientific Chemicals

Catalog number: L13396.06
5 g, Each
Thermo Scientific Chemicals

1-Pyrrolidinecarbonyl chloride, 97%, Thermo Scientific Chemicals

Catalog number: L13396.06
5 g, Each
Quantity
Catalog number: L13396.06
also known as L13396-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1192-63-8
IUPAC Name
pyrrolidine-1-carbonyl chloride
Molecular Formula
C5H8ClNO
InChI Key
XACWJIQLDLUFSR-UHFFFAOYSA-N
SMILES
ClC(=O)N1CCCC1
Identification (FTIR)
Conforms
Form
Liquid
Assay (GC)
≥96.0%
Appearance (Color)
Clear colorless to yellow
Refractive Index
1.4905-1.4955 @ 20?C

Description

1-Pyrrolidinecarbonyl chloride, is used in the synthesis of (S)-4-(2-(((benzyloxy)carbonyl)amino)-3-methoxy-3-oxopropyl)phenyl pyrrolidine-1-carboxylate

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Pyrrolidinecarbonyl chloride, is used in the synthesis of (S)-4-(2-(((benzyloxy)carbonyl)amino)-3-methoxy-3-oxopropyl)phenyl pyrrolidine-1-carboxylate

Solubility
It reacts with water.

Notes
Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents, bases. Stable under recommended storage conditions.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text