1,4-Cyclohexanebis(methylamine), cis + trans, 96%, Thermo Scientific Chemicals
1,4-Cyclohexanebis(methylamine), cis + trans, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1,4-Cyclohexanebis(methylamine), cis + trans, 96%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
25 g
Catalog number L13407.06
also known as L13407-06
Price (USD)/ Each
95.80
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
95.80
Add to cart
1,4-Cyclohexanebis(methylamine), cis + trans, 96%, Thermo Scientific Chemicals
Catalog numberL13407.06
Price (USD)/ Each
95.80
-
Add to cart
Chemical Identifiers
CAS2549-93-1
IUPAC Name[4-(azaniumylmethyl)cyclohexyl]methanaminium
Molecular FormulaC8H20N2
InChI KeyOXIKYYJDTWKERT-UHFFFAOYSA-P
SMILES[NH3+]CC1CCC(C[NH3+])CC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
FormLiquid
Assay (GC)≥95.0%
Refractive Index1.4890-1.4950 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only