Thermo Scientific Chemicals

Diethylene glycol monoethyl ether acetate, 99%, Thermo Scientific Chemicals

Catalog number: L13446.AP
500 mL, Each
Thermo Scientific Chemicals

Diethylene glycol monoethyl ether acetate, 99%, Thermo Scientific Chemicals

Catalog number: L13446.AP
500 mL, Each
Quantity
Catalog number: L13446.AP
also known as L13446-AP
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
112-15-2
Identification (FTIR)
Conforms
Water Content (Karl Fischer Titration)
≤0.5%
Refractive Index
1.4190-1.4225 @ 20?C
Appearance (Color)
Clear colorless
Form
Liquid

Description

Diethylene glycol monoethyl ether acetate is used as a catalytic agent, petrochemical additive, coating auxiliary agent and in organic synthesis. It is mainly used as metal, furniture paint solvents and as organic intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethylene glycol monoethyl ether acetate is used as a catalytic agent, petrochemical additive, coating auxiliary agent and in organic synthesis. It is mainly used as metal, furniture paint solvents and as organic intermediate.

Solubility
Soluble in water.

Notes
Store at room temperature. Incompatible with bases and oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers. Containers which are opened must be carefully resealed and kept upright to prevent leakage.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text