Thermo Scientific Chemicals

3,4,5-Trimethoxybenzylamine, 96%, Thermo Scientific Chemicals

Catalog number: L13461.14
25 g, Each
Thermo Scientific Chemicals

3,4,5-Trimethoxybenzylamine, 96%, Thermo Scientific Chemicals

Catalog number: L13461.14
25 g, Each
Quantity
Catalog number: L13461.14
also known as L13461-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
18638-99-8
IUPAC Name
1-(3,4,5-trimethoxyphenyl)methanamine
Molecular Formula
C10H15NO3
InChI Key
YUPUSBMJCFBHAP-UHFFFAOYSA-N
SMILES
COC1=CC(CN)=CC(OC)=C1OC
Assay (GC)
≥95.0%
Identification (FTIR)
Conforms
Refractive Index
1.5445-1.5505 @ 20?C
Appearance (Color)
Clear colorless to yellow
Form
Liquid or viscous liquid

Description

3,4,5-Trimethoxybenzylamine is used in the synthesis of benzyl 3-deoxy-3-(3,4,5-trimethoxybenzylamino)-β-L-xylopyranoside. It is a important organic intermediate. It is used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,4,5-Trimethoxybenzylamine is used in the synthesis of benzyl 3-deoxy-3-(3,4,5-trimethoxybenzylamino)-β-L-xylopyranoside. It is a important organic intermediate. It is used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Not miscible or difficult to mix in water.

Notes
It is sensitive to air. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text