Thermo Scientific Chemicals

2-Chloroaniline hydrochloride, 97%, Thermo Scientific Chemicals

Catalog number: L13483.22
100 g, Each
Thermo Scientific Chemicals

2-Chloroaniline hydrochloride, 97%, Thermo Scientific Chemicals

Catalog number: L13483.22
100 g, Each
Quantity
Catalog number: L13483.22
also known as L13483-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
137-04-2
IUPAC Name
hydrogen 2-chloroaniline chloride
Molecular Formula
C6H7Cl2N
InChI Key
DRGIDRZFKRLQTE-UHFFFAOYSA-N
SMILES
[H+].[Cl-].NC1=CC=CC=C1Cl
Appearance (Color)
White
Form
Crystals or powder or crystalline powder
Assay (Titration ex Chloride)
≥96.0 to ≤104.0%
Assay (GC)
≥96.0%

Description

2-Chloroaniline hydrochloride, is used as ice dye basement, used for towels, colored yarn and cloth of cotton, viscose fabric dyeing and printing of chromogenic agent. It is also used as an intermediate in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chloroaniline hydrochloride, is used as ice dye basement, used for towels, colored yarn and cloth of cotton, viscose fabric dyeing and printing of chromogenic agent. It is also used as an intermediate in organic synthesis.

Solubility
It is soluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text