Thermo Scientific Chemicals

4-Chloro-2-fluorobenzylamine hydrochloride, 97%, Thermo Scientific Chemicals

Catalog number: L13488.03
1 g, Each
Thermo Scientific Chemicals

4-Chloro-2-fluorobenzylamine hydrochloride, 97%, Thermo Scientific Chemicals

Catalog number: L13488.03
1 g, Each
Quantity
Catalog number: L13488.03
also known as L13488-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
202982-63-6
IUPAC Name
(4-chloro-2-fluorophenyl)methanaminium
Molecular Formula
C7H8ClFN
InChI Key
ODMMHGMIZYLHNN-UHFFFAOYSA-O
SMILES
[NH3+]CC1=CC=C(Cl)C=C1F
Assay (Titration ex Chloride)
≥96.0 to ≤104.0%
Appearance (Color)
White to cream
Form
Powder and/or lumps
Melting Point (clear melt)
239-247?C

Description

4-Chloro-2-fluorobenzylamine hydrochloride It is used as a primary and secondary intermediate. It is also used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Chloro-2-fluorobenzylamine hydrochloride It is used as a primary and secondary intermediate. It is also used as a pharmaceutical intermediate.

Solubility
It is soluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text