Thermo Scientific Chemicals

2-(Dimethylamino)ethyl acrylate, 98%, stab. with ca 0.1% 4-methoxyphenol, Thermo Scientific Chemicals

Catalog number: L13506.AC
25 mL, Each
Thermo Scientific Chemicals

2-(Dimethylamino)ethyl acrylate, 98%, stab. with ca 0.1% 4-methoxyphenol, Thermo Scientific Chemicals

Catalog number: L13506.AC
25 mL, Each
Quantity
Catalog number: L13506.AC
also known as L13506-AC
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2439-35-2
IUPAC Name
2-(dimethylamino)ethyl prop-2-enoate
Molecular Formula
C7H13NO2
InChI Key
DPBJAVGHACCNRL-UHFFFAOYSA-N
SMILES
CN(C)CCOC(=O)C=C
Appearance (Color)
Clear, colorless to yellow or pale brown
Assay (GC)
>97.5%
Refractive Index
1.4360-1.4400 @ 20?C
Solution Test
(10% v/v in diethyl ether) : Clear
Solution Test
(10% v/v in methanol) : Clear

Description

2-(Dimethylamino)ethyl acrylate is an important raw material and intermediate used in organic synthesis, pharmaceuticals, as a catalytic agent, as a petrochemical additive and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(Dimethylamino)ethyl acrylate is an important raw material and intermediate used in organic synthesis, pharmaceuticals, as a catalytic agent, as a petrochemical additive and agrochemicals.

Solubility
Soluble in water (1.000 g/L at 20°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Recommended storage temperature is 2 - 8°C. Sensitive to heat, light and moisture.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text