Thermo Scientific Chemicals

2-(Methacryloyloxy)ethyltrimethylammonium chloride, 72% aq. soln., stab. with 150-200 ppm 4-methoxyphenol, Thermo Scientific Chemicals

Catalog number: L13523.36
500 g, Each
Thermo Scientific Chemicals

2-(Methacryloyloxy)ethyltrimethylammonium chloride, 72% aq. soln., stab. with 150-200 ppm 4-methoxyphenol, Thermo Scientific Chemicals

Catalog number: L13523.36
500 g, Each
Quantity
Catalog number: L13523.36
also known as L13523-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5039-78-1
IUPAC Name
trimethyl({2-[(2-methylprop-2-enoyl)oxy]ethyl})azanium chloride
Molecular Formula
C9H18ClNO2
InChI Key
RRHXZLALVWBDKH-UHFFFAOYSA-M
SMILES
[Cl-].CC(=C)C(=O)OCC[N+](C)(C)C
Appearance (Color)
Clear colorless to pale yellow
Assay (HPLC)
≥67.0%
Assay (Titration ex Chloride)
≥66.5%
Form
Liquid
Refractive Index
1.4610-1.4730 @ 20°C

Description

2-(Methacryloyloxy)ethyltrimethylammonium chloride is a zirconium, aluminum and organic surface treated multipurpose rutile titanium dioxide pigment, it has narrow-size material distribution, good whiteness and high weather resistance. It is widely used in coating, plastics, paints, rubber and ink production, leather, cosmetics, soaps, plastics and in paper industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(Methacryloyloxy)ethyltrimethylammonium chloride is a zirconium, aluminum and organic surface treated multipurpose rutile titanium dioxide pigment, it has narrow-size material distribution, good whiteness and high weather resistance. It is widely used in coating, plastics, paints, rubber and ink production, leather, cosmetics, soaps, plastics and in paper industry.

Solubility
It is soluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text