Methyl 2-methoxypropionate, 98%, Thermo Scientific Chemicals
Methyl 2-methoxypropionate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Methyl 2-methoxypropionate, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L13527.06
also known as L13527-06
Price (USD)/ Each
44.80
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
44.80
Add to cart
Methyl 2-methoxypropionate, 98%, Thermo Scientific Chemicals
Catalog numberL13527.06
Price (USD)/ Each
44.80
-
Add to cart
Chemical Identifiers
CAS17639-76-8
IUPAC Namemethyl 2-methoxypropanoate
Molecular FormulaC5H10O3
InChI KeyVABBJJOSOCPYIT-UHFFFAOYNA-N
SMILESCOC(C)C(=O)OC
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5%
Refractive Index1.3945-1.3985 @ 20?C
Appearance (Color)Clear colorless to pale yellow
FormLiquid
Methyl 2-methoxypropionate, is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 2-methoxypropionate, is used as a pharmaceutical intermediate.

Solubility
Soluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
RUO – Research Use Only

General References:

  1. H Kanai.; K Ishii. Hydrodimerization of methyl acrylate catalyzed by halogenotris (triphenylphosphine) cobalt. Bulletin of the Chemical Society of Japan. 1981, 54,(4), 1015-1018.
  2. WA Bonner.; JA Zderic. The Stereochemistry of Raney Nickel Action. VI. Functional Group Hydrogenolyses in the α-Substituted α-Phenylpropionic Acid Series. Journal of the American Chemical Society. 1956, 78,(13), 3218-3221.